Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027583.3 | NP_081859.1 | 7 - 8 | 1351 | 111 | |
NM_029150.4 | NP_083426.1 | 7 - 8 | 1351 | 111 | ||
GenBank mRNA | AK014869.1 | - | 7 - 8 | 1315 | 111 | |
AK076628.1 | - | 7 - 8 | 1352 | 111 | ||
BC100443.1 | - | 7 - 8 | 1332 | 30920480 | 111 | |
BC132205.1 | - | 7 - 8 | 1275 | 40130482 | 111 | |
BC132539.1 | - | 7 - 8 | 1275 | 40130816 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |