Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF037205.1 | - | 4 - 5 | 410 | 85 | |
AF037206.1 | - | 4 - 5 | 410 | 85 | ||
AK034135.1 | - | 4 - 5 | 461 | 85 | ||
AK036591.1 | - | 4 - 5 | 455 | 85 | ||
AK050095.1 | - | 4 - 5 | 423 | 85 | ||
AK082296.1 | - | 4 - 5 | 451 | 85 | ||
AK139447.1 | - | 4 - 5 | 416 | 85 | ||
AK140682.1 | - | 4 - 5 | 465 | 85 | ||
AK158046.1 | - | 4 - 5 | 451 | 85 | ||
AK167278.1 | - | 4 - 5 | 415 | 85 | ||
BC058182.1 | - | 4 - 5 | 418 | 5324040 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |