Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144987.1 | NP_001138459.1 | 7 - 8 | 1982 | 111 | |
NM_001144988.1 | NP_001138460.1 | 6 - 7 | 1595 | 111 | ||
NM_144895.2 | NP_659144.1 | 7 - 8 | 1766 | 111 | ||
GenBank mRNA | AK031545.1 | - | 6 - 7 | 1321 | 111 | |
AK035081.1 | - | 5 - 6 | 1454 | 111 | ||
AK036569.1 | - | 5 - 6 | 918 | 111 | ||
AK087388.1 | - | 7 - 8 | 1735 | 111 | ||
AK089946.1 | - | 7 - 8 | 1767 | 111 | ||
AK129171.1 | - | 6 - 7 | 1574 | 111 | ||
AK147760.1 | - | 7 - 8 | 1983 | 111 | ||
AK159951.1 | - | 7 - 8 | 1767 | 111 | ||
BC022921.1 | - | 7 - 8 | 1734 | 4220506 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |