Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027866.1 | NP_082142.1 | 3 - 4 | 284 | 66 | |
GenBank mRNA | AK003121.1 | - | 3 - 4 | 284 | 66 | |
BC104380.1 | - | 3 - 4 | 255 | 40050136 | 66 | |
BC104381.1 | - | 3 - 4 | 255 | 40050143 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |