Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm19835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138371.1 | - | 71 | 100 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sosondowah ankyrin repeat domain family member A | Sowaha |
A830006N08Rik, Ankrd43 |
-2.9kb | Sense | Chr. 11 - 53476578 - 53480195 on NCBI Build GRCm38 |
Downstream | septin 8 | Sept8 |
AW046166, Sepl, mKIAA0202 |
32.4kb | Anti-sense | Chr. 11 - 53519736 - 53549600 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons |