Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300007104.mm.1 |
NM_001271388.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007104.mm.1 |
NM_001271390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007104.mm.1 |
NM_001271391.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007104.mm.1 |
NM_001271392.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007104.mm.1 |
NM_023472.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007104.mm.1 |
TC1300001112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300007103.mm.1 |
NM_001271388.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007103.mm.1 |
NM_001271390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007103.mm.1 |
NM_001271391.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007103.mm.1 |
NM_001271392.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007103.mm.1 |
NM_023472.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300007103.mm.1 |
TC1300001112.mm.1 |
MTA TC |