Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008429.2 | NP_032455.2 | 1 - 2 | 130 | 60 | |
GenBank mRNA | AF130860.1 | - | 1 - 2 | 93 | 60 | |
AK033970.1 | - | 1 - 2 | 130 | 60 | ||
AK046084.1 | - | 1 - 2 | 169 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |