Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028057.2 | NP_082333.1 | 4 - 5 | 386 | 99 | |
GenBank mRNA | AK005159.1 | - | 4 - 5 | 387 | 99 | |
AK049717.1 | - | 3 - 4 | 608 | 99 | ||
AK159663.1 | - | 4 - 5 | 356 | 99 | ||
AK167178.1 | - | 4 - 5 | 380 | 99 | ||
BC016266.1 | - | 4 - 5 | 595 | 4924006 | 99 | |
BC024618.1 | - | 4 - 5 | 364 | 4235599 | 99 | |
JX420288.1 | - | 466 | 99 | |||
JX420290.1 | - | 4 - 5 | 563 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |