Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599497 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599491 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599492 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599495 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599490 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599489 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599488 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599493 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
BY631266.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
AK147795.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
AK129098.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
NM_026390.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
BY631266.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
AK147795.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
AK129098.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P401475 |
NM_026390.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
937873 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
526506 |
AK014416.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
526506 |
NM_027510.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
494042 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777215 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431711 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
46747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
423594 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895163 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
732625 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
813549 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
192792 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
298175 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
770966 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
410791 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
357874 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580076 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
473065 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
452995 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
892468 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
915211 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140899 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54117 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
137670 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
23913 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
46290 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P104742 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P180192 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
BY631266.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
AK147795.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
AK129098.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
NM_026390.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
BY631266.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
AK147795.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
AK129098.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P401475 |
NM_026390.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599483 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599491 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599498 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599505 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599481 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599475 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599464 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599465 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599471 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599500 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599478 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599502 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599467 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599497 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599479 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599472 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599490 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599503 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599504 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599487 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599484 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599468 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599485 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599494 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599473 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20599493 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P180192 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04257265 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P104742 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04257247 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P184312 |
BC012271.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P170610 |
BC019795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P170610 |
AK147795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P170610 |
AK148889.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P170610 |
AK129098.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P170610 |
AK145551.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P298145 |
BC012271.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P298145 |
BC019795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P298145 |
BC030190.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P298145 |
AK147795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P298145 |
AK148333.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
BY631266.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
AK147795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
AK129098.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
NM_026390.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
BY631266.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
AK147795.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
AK129098.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P401475 |
NM_026390.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130402 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P104742 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P180192 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00516477 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00516479 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01125928 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00516476 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01125932 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599489 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599495 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599492 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599490 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599488 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599497 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599491 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20599493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599563 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599580 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20599464 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599508 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599464 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599476 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599580 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599518 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599513 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599500 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599467 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599507 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599477 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00914569 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599505 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599472 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599475 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599471 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599498 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599479 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599514 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599502 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00917935 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599512 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599490 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00534726 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00907040 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599582 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599577 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00516476 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599584 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00023577 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599527 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599516 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599465 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00917937 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599497 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00534719 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599484 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599581 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599526 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599578 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00306448 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599583 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599504 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20599493 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00917933 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00027553 |
- |
- |