Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159717.1 | NP_001153189.1 | 5 - 6 | 431 | 99 | |
NM_001159718.1 | NP_001153190.1 | 4 - 5 | 404 | 99 | ||
NM_001159719.1 | NP_001153191.1 | 5 - 6 | 479 | 99 | ||
NM_010891.2 | NP_035021.1 | 5 - 6 | 435 | 99 | ||
GenBank mRNA | AK028072.1 | - | 5 - 6 | 417 | 99 | |
AK146616.1 | - | 5 - 6 | 432 | 99 | ||
AK151591.1 | - | 5 - 6 | 419 | 99 | ||
AK169852.1 | - | 4 - 5 | 454 | 99 | ||
AK171331.1 | - | 5 - 6 | 414 | 99 | ||
BC138636.1 | - | 4 - 5 | 372 | 8861656 | 99 | |
BC138637.1 | - | 4 - 5 | 372 | 8861657 | 99 | |
D49382.2 | - | 5 - 6 | 390 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |