Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133805.3 | NP_598566.3 | 2 - 3 | 271 | 87 | |
GenBank mRNA | AF482000.1 | - | 2 - 3 | 732 | 87 | |
AK079111.1 | - | 2 - 3 | 205 | 87 | ||
AK079522.1 | - | 2 - 3 | 219 | 87 | ||
AK088323.1 | - | 2 - 3 | 191 | 87 | ||
AK090013.1 | - | 2 - 3 | 200 | 87 | ||
AK145899.1 | - | 2 - 3 | 198 | 87 | ||
AK162502.1 | - | 2 - 3 | 206 | 87 | ||
AK195692.1 | - | 1 - 2 | 46 | 87 | ||
BC017690.1 | - | 2 - 3 | 239 | 4238947 | 87 | |
BC021488.1 | - | 2 - 3 | 230 | 5344864 | 87 | |
BC024421.1 | - | 2 - 3 | 224 | 4987584 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |