Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013919.4 | NP_038947.2 | 1 - 2 | 201 | 104 | |
GenBank mRNA | AK013252.1 | - | 1 - 2 | 196 | 104 | |
BC021903.1 | - | 1 - 2 | 191 | 3985792 | 104 | |
BY110597.1 | - | 1 - 2 | 201 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |