Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077189.1 | NP_001070657.1 | 6 - 7 | 970 | 58 | |
GenBank mRNA | AK155329.1 | - | 6 - 7 | 58 | ||
AK171254.1 | - | 6 - 7 | 58 | |||
AK171316.1 | - | 6 - 7 | 953 | 58 | ||
M16367.1 | - | 8 - 9 | 1208 | 58 | ||
U31801.1 | - | 4 - 5 | 785 | 58 | ||
U31802.1 | - | 4 - 5 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |