Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026250.2 | NP_080526.2 | 5 - 6 | 916 | 82 | |
GenBank mRNA | AK015243.1 | - | 5 - 6 | 895 | 82 | |
AK090002.1 | - | 5 - 6 | 634 | 82 | ||
AK169361.1 | - | 5 - 6 | 879 | 82 | ||
BC033572.1 | - | 4 - 5 | 861 | 5067692 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |