Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178079.3 | NP_835180.2 | 4 - 5 | 604 | 103 | |
GenBank mRNA | AK028883.1 | - | 4 - 5 | 601 | 103 | |
AK128969.1 | - | 4 - 5 | 610 | 103 | ||
AK143953.1 | - | 4 - 5 | 599 | 103 | ||
BC025830.1 | - | 1 - 2 | 94 | 103 | ||
BC120725.1 | - | 4 - 5 | 604 | 40129648 | 103 | |
BC125395.1 | - | 4 - 5 | 604 | 40129910 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |