Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178079.3 | NP_835180.2 | 5 - 6 | 729 | 62 | |
GenBank mRNA | AK028883.1 | - | 5 - 6 | 726 | 62 | |
AK128969.1 | - | 5 - 6 | 735 | 62 | ||
AK143953.1 | - | 5 - 6 | 724 | 62 | ||
BC025830.1 | - | 2 - 3 | 219 | 62 | ||
BC120725.1 | - | 5 - 6 | 729 | 40129648 | 62 | |
BC125395.1 | - | 5 - 6 | 729 | 40129910 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |