Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081756.1 | NP_001075225.1 | 4 - 5 | 579 | 67 | |
NM_172484.3 | NP_766072.2 | 2 - 3 | 440 | 67 | ||
NM_176957.3 | NP_795931.2 | 4 - 5 | 579 | 67 | ||
GenBank mRNA | AK053243.1 | - | 2 - 3 | 262 | 67 | |
BC066095.1 | - | 2 - 3 | 440 | 6856184 | 67 | |
BC079551.1 | - | 2 - 3 | 440 | 6856184 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |