Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172157.1 | NP_001165628.1 | 14 - 15 | 1863 | 74 | |
NM_153530.2 | NP_705758.1 | 13 - 14 | 1821 | 74 | ||
GenBank mRNA | AK029652.1 | - | 14 - 15 | 1877 | 74 | |
AK031180.1 | - | 14 - 15 | 1846 | 74 | ||
BC036177.1 | - | 13 - 14 | 1782 | 5004341 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |