Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172862.3 | NP_766450.2 | 20 - 21 | 8783 | 82 | |
GenBank mRNA | AB236662.1 | - | 20 - 21 | 8783 | 82 | |
AJ833643.1 | - | 20 - 21 | 8512 | 82 | ||
AK082256.1 | - | 3 - 4 | 813 | 82 | ||
BC068185.1 | - | 10 - 11 | 1685 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |