Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183221.3 | NP_899044.3 | 1 - 2 | 5195 | 99 | |
GenBank mRNA | AK034552.1 | - | 1 - 2 | 1193 | 99 | |
DQ286572.1 | - | 1 - 2 | 5195 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |