Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033409.3 | NP_001028581.1 | 8 - 9 | 991 | 85 | |
GenBank mRNA | AK085901.1 | - | 8 - 9 | 949 | 85 | |
AK137380.1 | - | 8 - 9 | 992 | 85 | ||
BC141210.1 | - | 8 - 9 | 943 | 9055822 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |