Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253708.1 | NP_001240637.1 | 6 - 7 | 2009 | 62 | |
NM_001253709.1 | NP_001240638.1 | 5 - 6 | 855 | 62 | ||
NM_001253710.1 | NP_001240639.1 | 5 - 6 | 960 | 62 | ||
NM_001253713.1 | NP_001240642.1 | 6 - 7 | 963 | 62 | ||
NM_020007.3 | NP_064391.2 | 6 - 7 | 2009 | 62 | ||
GenBank mRNA | AB093240.1 | - | 6 - 7 | 2013 | 62 | |
AF231110.1 | - | 5 - 6 | 1449 | 62 | ||
AK153827.1 | - | 5 - 6 | 853 | 62 | ||
AK153848.1 | - | 5 - 6 | 843 | 62 | ||
BC096600.1 | - | 5 - 6 | 921 | 4976244 | 62 | |
CN663545.1 | - | 3 - 4 | 536 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |