Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207531.3 | NP_997414.2 | 6 - 7 | 500 | 62 | |
GenBank mRNA | AK053177.1 | - | 6 - 7 | 429 | 62 | |
AW490945.1 | - | 2 - 3 | 62 | |||
BC023499.1 | - | 6 - 7 | 501 | 5008294 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |