Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146033.3 | NP_666145.3 | 2 - 3 | 368 | 94 | |
GenBank mRNA | AK077556.1 | - | 2 - 3 | 339 | 94 | |
AK164434.1 | - | 2 - 3 | 270 | 94 | ||
BC012275.1 | - | 2 - 3 | 334 | 4010781 | 94 | |
BC024959.1 | - | 2 - 3 | 325 | 4946879 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |