Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021537.3 | NP_067512.3 | 7 - 8 | 1008 | 90 | |
GenBank mRNA | AF004934.1 | - | 6 - 7 | 846 | 90 | |
AK148041.1 | - | 7 - 8 | 982 | 90 | ||
BC029875.1 | - | 6 - 7 | 1726 | 90 | ||
BC052913.1 | - | 7 - 8 | 943 | 30057101 | 90 | |
BC071218.1 | - | 7 - 8 | 1003 | 4012695 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |