Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026390.2 | NP_080666.2 | 3 - 4 | 505 | 82 | |
GenBank mRNA | AK011233.1 | - | 3 - 4 | 402 | 82 | |
AK012824.1 | - | 3 - 4 | 399 | 82 | ||
AK019371.1 | - | 3 - 4 | 398 | 82 | ||
AK029431.1 | - | 3 - 4 | 551 | 82 | ||
AK032206.1 | - | 3 - 4 | 384 | 82 | ||
AK147795.1 | - | 3 - 4 | 378 | 82 | ||
AK148333.1 | - | 3 - 4 | 391 | 82 | ||
BC012271.1 | - | 3 - 4 | 517 | 82 | ||
BC019795.1 | - | 3 - 4 | 524 | 4158848 | 82 | |
BC030190.1 | - | 3 - 4 | 514 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |