Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159745.1 | NP_001153217.1 | 1 - 2 | 451 | 92 | |
NM_009183.2 | NP_033209.1 | 1 - 2 | 451 | 92 | ||
GenBank mRNA | AJ223956.1 | - | 1 - 2 | 442 | 92 | |
AK003690.1 | - | 1 - 2 | 393 | 92 | ||
AK041723.1 | - | 1 - 2 | 384 | 92 | ||
AK165717.1 | - | 1 - 2 | 426 | 92 | ||
BC060112.1 | - | 1 - 2 | 428 | 6405078 | 92 | |
X86000.1 | - | 451 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |