Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145991.1 | NP_666103.1 | 4 - 5 | 431 | 113 | |
GenBank mRNA | AK080861.1 | - | 4 - 5 | 563 | 113 | |
BC027756.1 | - | 4 - 5 | 532 | 5067496 | 113 | |
BC031127.1 | - | 4 - 5 | 431 | 4972826 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |