Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctage5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165253.1 | NP_001158725.1 | 6 - 7 | 584 | 129 | |
NM_001165254.1 | NP_001158726.1 | 6 - 7 | 583 | 129 | ||
NM_146034.3 | NP_666146.3 | 8 - 9 | 682 | 129 | ||
GenBank mRNA | AK029466.1 | - | 6 - 7 | 583 | 129 | |
AK053010.1 | - | 7 - 8 | 589 | 129 | ||
AK080312.1 | - | 7 - 8 | 631 | 129 | ||
AK087188.1 | - | 7 - 8 | 597 | 129 | ||
AK157358.1 | - | 6 - 7 | 585 | 129 | ||
BC026864.1 | - | 8 - 9 | 634 | 2647214 | 129 | |
BQ571563.1 | - | 6 - 7 | 583 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |