Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009147.2 | NP_033173.2 | 12 - 13 | 1596 | 77 | |
GenBank mRNA | AK028152.1 | - | 12 - 13 | 1484 | 77 | |
AY082671.1 | - | 12 - 13 | 1470 | 77 | ||
BC034610.1 | - | 12 - 13 | 1596 | 5364796 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |