Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009147.2 | NP_033173.2 | 17 - 18 | 2181 | 88 | |
GenBank mRNA | AK028152.1 | - | 17 - 18 | 2069 | 88 | |
AY082671.1 | - | 17 - 18 | 2055 | 88 | ||
BC034610.1 | - | 17 - 18 | 2181 | 5364796 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |