Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081281.1 | NP_001074750.1 | 3 - 4 | 658 | 66 | |
GenBank mRNA | AK052918.1 | - | 3 - 4 | 659 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |