Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081125.1 | NP_001074594.1 | 5 - 6 | 976 | 63 | |
GenBank mRNA | AM261669.1 | - | 2 - 3 | 220 | 63 | |
AM262742.1 | - | 4 - 5 | 493 | 63 | ||
AM262743.1 | - | 4 - 5 | 590 | 63 | ||
AM262744.1 | - | 6 - 7 | 807 | 63 | ||
BC082604.1 | - | 5 - 6 | 917 | 30672479 | 63 | |
BC085190.1 | - | 5 - 6 | 892 | 30694633 | 63 | |
X99104.1 | - | 5 - 6 | 736 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |