Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023755.2 | NP_076244.2 | 10 - 11 | 1095 | 71 | |
GenBank mRNA | AF311309.1 | - | 10 - 11 | 1096 | 71 | |
AK143960.1 | - | 10 - 11 | 1094 | 71 | ||
BC116663.1 | - | 10 - 11 | 1090 | 40087602 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |