Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 24 - 25 | 4471 | 79 | |
GenBank mRNA | AF300701.1 | - | 23 - 24 | 3948 | 79 | |
AK156112.1 | - | 24 - 25 | 4474 | 79 | ||
U36488.1 | - | 20 - 21 | 3633 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mus musculus Probe spans exons |