Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 35 - 36 | 5752 | 112 | |
GenBank mRNA | AF300701.1 | - | 34 - 35 | 5229 | 112 | |
AK156112.1 | - | 35 - 36 | 5755 | 112 | ||
U36488.1 | - | 31 - 32 | 4914 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Detect Genomic DNA Mus musculus Probe spans exons |