Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00003974.mm.1 |
NM_001081354.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003974.mm.1 |
NM_001081354.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003974.mm.1 |
NM_001256048.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003974.mm.1 |
NM_001256048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003974.mm.1 |
TC0X00000643.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003976.mm.1 |
NM_001081354.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003976.mm.1 |
NM_001081354.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003976.mm.1 |
NM_001256048.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003976.mm.1 |
NM_001256048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003976.mm.1 |
TC0X00000643.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003977.mm.1 |
NM_001081354.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003977.mm.1 |
NM_001081354.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003977.mm.1 |
NM_001256048.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003977.mm.1 |
NM_001256048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003977.mm.1 |
TC0X00000643.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003975.mm.1 |
NM_001081354.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003975.mm.1 |
NM_001081354.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003975.mm.1 |
NM_001256048.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003975.mm.1 |
NM_001256048.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003975.mm.1 |
TC0X00000643.mm.1 |
MTA TC |