Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177330.4 | NP_796304.1 | 1 - 2 | 88 | 95 | |
GenBank mRNA | AK049671.1 | - | 1 - 2 | 89 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Mus musculus Probe spans exons |