Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173772.3 | NP_776133.1 | 1 - 2 | 123 | 64 | |
GenBank mRNA | AK034236.1 | - | 1 - 2 | 94 | 64 | |
BC070438.1 | - | 1 - 2 | 123 | 6842155 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |