Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173772.3 | NP_776133.1 | 2 - 3 | 357 | 105 | |
GenBank mRNA | AB510404.1 | - | 3 - 4 | 338 | 105 | |
AK034236.1 | - | 2 - 3 | 328 | 105 | ||
AY258421.1 | - | 2 - 3 | 270 | 105 | ||
BC070438.1 | - | 2 - 3 | 357 | 6842155 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |