Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153393.2 | NP_700442.2 | 25 - 26 | 1628 | 68 | |
GenBank mRNA | AF410792.1 | - | 25 - 26 | 1628 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |