Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011439.2 | NP_035569.2 | 1 - 2 | 342 | 81 | |
GenBank mRNA | BC007130.1 | - | 1 - 2 | 164 | 3493465 | 81 |
BE949927.1 | - | 1 - 2 | 342 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |