Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026164.2 | NP_080440.2 | 3 - 4 | 1419 | 132 | |
GenBank mRNA | AB044139.1 | - | 3 - 4 | 1298 | 132 | |
AK004621.1 | - | 3 - 4 | 1386 | 132 | ||
AK084928.1 | - | 1086 | 132 | |||
AK145776.1 | - | 3 - 4 | 1329 | 132 | ||
AK163211.1 | - | 3 - 4 | 1387 | 132 | ||
AK168475.1 | - | 3 - 4 | 1382 | 132 | ||
BC127056.1 | - | 2 - 3 | 1279 | 40109753 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |