Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193266.1 | NP_001180195.2 | 8 - 9 | 3120 | 97 | |
NM_207010.2 | NP_996893.3 | 8 - 9 | 3120 | 97 | ||
GenBank mRNA | AK137667.1 | - | 8 - 9 | 3121 | 97 | |
AY371925.1 | - | 8 - 9 | 1774 | 97 | ||
BC137901.1 | - | 8 - 9 | 1773 | 8860920 | 97 | |
BC137902.1 | - | 8 - 9 | 1773 | 8860921 | 97 | |
BC145021.1 | - | 8 - 9 | 1768 | 97 | ||
BC171938.1 | - | 8 - 9 | 1773 | 9054224 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |