Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076941 |
BC117055.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076941 |
NM_001081406.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076941 |
AK010419.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076941 |
AK144741.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2076941 |
BC022648.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
BC117055.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
NM_001081406.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
AK010419.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
AK144741.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
BC022648.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
BC117055.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
NM_001081406.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
AK010419.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
AK144741.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P347240 |
BC022648.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376401 |
AK003192.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376401 |
AK003286.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376401 |
AK160697.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376401 |
BC028523.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376401 |
BC079614.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376401 |
NM_026729.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156259 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
919843 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
641176 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520321 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
964223 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
728693 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151216 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P172182 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P161754 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076941 |
BC117055.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076941 |
NM_001081406.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076941 |
AK010419.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076941 |
AK144741.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2076941 |
BC022648.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
BC117055.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
NM_001081406.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
AK010419.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
AK144741.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
BC022648.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
BC117055.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
NM_001081406.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
AK010419.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
AK144741.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P347240 |
BC022648.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07653691 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P172182 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P161754 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P347240 |
BC117055.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
NM_001081406.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
AK010419.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
AK144741.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
BC022648.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
BC117055.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
NM_001081406.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
AK010419.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
AK144741.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347240 |
BC022648.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P161754 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P172182 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10318018 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10318007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513248 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513137 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513228 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28513149 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10363554 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513139 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513156 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513199 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513140 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513128 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513141 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513181 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513165 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513146 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513143 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10285267 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513188 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10318015 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513147 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513201 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513172 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513197 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513160 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10159390 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513136 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513171 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513138 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09809863 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513134 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513130 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09753128 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513195 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513168 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513170 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513185 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513186 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513192 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10191849 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513175 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513154 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09777763 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513162 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513183 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513176 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513174 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10254021 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513152 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513158 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513190 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28513194 |
- |
- |