Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175334.3 | NP_780543.2 | 2 - 3 | 2029 | 60 | |
GenBank mRNA | AK086146.1 | - | 1 - 2 | 1276 | 60 | |
AK129088.1 | - | 2 - 3 | 2009 | 60 | ||
AY442299.1 | - | 2 - 3 | 1986 | 60 | ||
BC058658.1 | - | 1 - 2 | 1369 | 60 | ||
BC059270.1 | - | 2 - 3 | 1497 | 60 | ||
BC094599.1 | - | 1 - 2 | 1227 | 6407060 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |