Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tstd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164525.1 | NP_001157997.1 | 1 - 2 | 102 | 56 | |
GenBank mRNA | BY720852.1 | - | 1 - 2 | 71 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |