Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010498.3 | NP_034628.2 | 3 - 4 | 608 | 82 | |
GenBank mRNA | AJ627044.1 | - | 1 - 2 | 73 | 82 | |
AK085237.1 | - | 4 - 5 | 485 | 82 | ||
AK132287.1 | - | 4 - 5 | 488 | 82 | ||
AK166178.1 | - | 3 - 4 | 486 | 82 | ||
AK166443.1 | - | 4 - 5 | 437 | 82 | ||
L07921.1 | - | 3 - 4 | 485 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |