Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013779.2 | NP_001013801.2 | 2 - 3 | 220 | 118 | |
GenBank mRNA | AK050724.1 | - | 2 - 3 | 220 | 118 | |
BC033443.1 | - | 2 - 3 | 226 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |