Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009190.2 | NP_033216.2 | 7 - 8 | 1065 | 76 | |
GenBank mRNA | AK151023.1 | - | 7 - 8 | 1019 | 76 | |
AK152131.1 | - | 7 - 8 | 1052 | 76 | ||
AK165594.1 | - | 1010 | 76 | |||
AK168270.1 | - | 7 - 8 | 956 | 76 | ||
BC003799.1 | - | 7 - 8 | 972 | 3488471 | 76 | |
U10119.1 | - | 7 - 8 | 1016 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |